missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRANDTECH™ BRAND™ Calibration Tool for Dispensette™ S Bottltop Dispensers (All Models)
Description
- For Dispensette™ S, Dispensette™ S Organic and Dispensette™ S Trace Analysis bottletop dispensers made 2106 and later.
- One end is used to mount and remove filling and discharge valves
- Other end is used to perform calibration adjustments on analog and fixed volume models
- Can also be used to assist of removal of back housing on digital models
Specifications
Specifications
| Product Type | Mounting Tool |
| Autoclavable | Autoclavable |
| Color | Black |
| Material | Polypropylene |
| For Use With (Equipment) | Dispensette S |
| Includes | Tool only |