missing translation for 'onlineSavingsMsg'
Learn More

3M™ Calibration Kit, 529-04-49

Catalog No. 296358264
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
296358264 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 296358264 Supplier 3M™ Supplier No. 7000131704

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used to calibrate the carbon monoxide (CO) monitors in the 3M™ Portable Compressed Air Filter and Regulator Panel 256-03-00, 3M™ Portable Compressed Air Filter and Regulator Panel, 256-02-01 and 3M™ Retrofit CO Monitor Kit, W-2808.

  • Delve into 3M™ Products and experience the wide variety of products and replacement parts offered with powered air purifying respirators (PAPR)
  • In addition to powered systems, 3M also offers supplied air respirators (SAR) using air from an air tank or a different external source

Specifications

Color Black
Applications Assembly, Blasting, Cleaning, Metal Repair, Painting
Quantity 1 Ea.
Product Type Calibration Kit