Learn More
BINDER™ Calibration Certificate
Catalog No. 08101248
Change view
Click to view available options
Calibration Points:
1 (Center of Chamber)
1 (Extension of Measurement)
15 to 18 Measuring Points
27 Measuring Points
9 Measuring Points
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Calibration Points
08101248 1 (Extension of Measurement)
08101244 1 (Center of Chamber)
08101245 15 to 18 Measuring Points
08101246 27 Measuring Points
08101247 9 Measuring Points
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 08101248 Supplier BINDER™ Supplier No. 80122231
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The calibration certificate includes temperature measurement at different measuring points or set values for the Series FP Avantgarde.Line Drying and Heating Chambers.
  • Measurement at specified temperature
  • Including certificate
  • Various points of measurement
  • Accessory for Series FP Avantgarde.Line Drying and Heating Chambers
Specifications
For Use With (Equipment) Series FP Avantgarde.Line Drying and Heating Chambers
Calibration Points 1 (Extension of Measurement)
Duration 1 Year
Product Type Calibration Certificate (Temperature)
Product Line BINDER™ FP Avantgarde.Line
Service Offering Operational Qualification
Includes Extension of measurement in center of chamber