missing translation for 'onlineSavingsMsg'
Learn More

Calcium tungsten oxide, 98%

CAS: 7790-75-2

$77.51 - $601.97

Chemical Identifiers

CAS 7790-75-2
MDL Number MFCD00010913
Synonym Calcium tungstate
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1305114
View Documents
Thermo Scientific Chemicals
01305114
25 g N/A
Only null left
 
AA1305122
View Documents
Thermo Scientific Chemicals
01305122
100 g N/A
Only null left
 
AA1305136
View Documents
Thermo Scientific Chemicals
01305136
500 g N/A
Only null left
 
Description

Description

Calcium tungsten oxide is used as a luminophore.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7790-75-2
MFCD00010913
Calcium tungstate

Specifications

1620°C
Powder
MFCD00010913
Calcium tungstate
287.93
Calcium tungsten oxide
7790-75-2
25 g
14,1712
Insoluble in H2O; Decomposes by hot HCl,HNO3
98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 232-219-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only