missing translation for 'onlineSavingsMsg'
Learn More

Calcium trifluoromethanesulfonate, 99% min

Catalog No. AA4013422
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4013422 100 g
AA4013409 10 g
AA4013418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4013422 Supplier Thermo Scientific Chemicals Supplier No. 04013422
Only null left

CAS: 55120-75-7 | C2CaF6O6S2 | 338.204 g/mol

Calcium trifluoromethanesulfonate is used for aminolysis of epoxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55120-75-7
Molecular Formula C2CaF6O6S2
Molecular Weight (g/mol) 338.204
MDL Number MFCD00143911
InChI Key PUQLFUHLKNBKQQ-UHFFFAOYSA-L
Synonym calcium trifluoromethanesulfonate, calcium triflate, methanesulfonic acid, trifluoro-, calcium salt, calcium trifluoromethansulphonate, methanesulfonic acid, 1,1,1-trifluoro-, calcium salt 2:1, calcium ditriflate, calcium trifluoromethyl sulfonate, calcium ii triflate, calcium 2+ ditriflate
PubChem CID 2734307
IUPAC Name calcium;trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Ca+2]

Specifications

Melting Point (decomposition)
Color White
Physical Form Powder
Quantity 100 g
Linear Formula Ca(OSO2CF3)2
Solubility Information Soluble in water.
Formula Weight 338.21
Percent Purity ≤99%
Chemical Name or Material Calcium trifluoromethanesulfonate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 232-188-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only