missing translation for 'onlineSavingsMsg'
Learn More

Calcium titanium oxide, 99+% (metals basis)

Catalog No. p-4781840
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1139736 500 g
AA11397A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1139736 Supplier Thermo Scientific Chemicals Supplier No. 01139736

CAS: 12049-50-2 | CaO3Ti | 135.94 g/mol

Calcium titanium oxide is used to prepare the ferrotitanium alloys and its structure is used to study the other compounds such as barium titanate.It is used to make the pure substances at low temperature.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12049-50-2
Molecular Formula CaO3Ti
Molecular Weight (g/mol) 135.94
MDL Number MFCD00015981
InChI Key WEUCVIBPSSMHJG-UHFFFAOYSA-N
Synonym calcium titanate, titaniumcalciumoxide, calciumtitaniumtrioxide, ca.o3ti, titanatecalcium 1:1, calcium dioxido oxo titanium, calcium oxotitaniumbis olate, calcium 2+ oxotitaniumbis olate, titanate tio32-,calcium 1:1, calcium titanate, nanopowder, <100 nm particle size bet trace metals basis
PubChem CID 16212381
IUPAC Name calcium;dioxido(oxo)titanium
SMILES [O--].[O--].[O--].[Ca++].[Ti+4]

Specifications

Melting Point 1975°C
Color Beige
Physical Form Powder
Quantity 500 g
Assay Percent Range >99% (Metals basis)
Solubility Information Insoluble in water. Soluble in oirganic and inorganic solution.
Formula Weight 135.98
Odor Odorless
Density 4.10 g/cm3
Chemical Name or Material Calcium titanium oxide
EINECSNumber 234-988-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only