missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium titanium oxide, 99+% (metals basis)
Description
Calcium titanium oxide is used to prepare the ferrotitanium alloys and its structure is used to study the other compounds such as barium titanate.It is used to make the pure substances at low temperature.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1975°C |
| Color | Beige |
| Physical Form | Powder |
| Quantity | 500 g |
| Odor | Odorless |
| Assay Percent Range | >99% (Metals basis) |
| Density | 4.10 g/cm3 |
| Solubility Information | Insoluble in water. Soluble in oirganic and inorganic solution. |
| Formula Weight | 135.98 |
| Chemical Name or Material | Calcium titanium oxide |
Safety and Handling
| EINECSNumber | 234-988-1 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only