missing translation for 'onlineSavingsMsg'
Learn More

Calcium tetrafluoroborate hydrate

Catalog No. p-7049829
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3013114 25 g
AA3013122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3013114 Supplier Thermo Scientific Chemicals Supplier No. 03013114
Only null left

CAS: 206996-55-6 | B2CaF8 | 213.685 g/mol

Calcium tetrafluoroborate hydrate is used as pharmaceutical and chemical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 206996-55-6
Molecular Formula B2CaF8
Molecular Weight (g/mol) 213.685
MDL Number MFCD00015972
InChI Key YHLNCCJTKRTQEG-UHFFFAOYSA-N
Synonym calcium tetrafluoroborate, calcium ditetrafluoroborate, calcium tetrafluoroborate 1-, acmc-1bo7f
PubChem CID 6096284
IUPAC Name calcium;ditetrafluoroborate
SMILES [B-](F)(F)(F)F.[B-](F)(F)(F)F.[Ca+2]

Specifications

Physical Form Powder
Quantity 25 g
Solubility Information Soluble in water.
Formula Weight 213.68 (Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Calcium tetrafluoroborate hydrate
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 237-485-5
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only