missing translation for 'onlineSavingsMsg'
Learn More

Calcium Sulfite, MP Biomedicals

Catalog No. ICN207810
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN207810 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN207810 Supplier MP Biomedicals Inc Supplier No. 0520781080

Chemical Identifiers

CAS 10257-55-3
Molecular Formula CaH2O3S
Molecular Weight (g/mol) 122.15
MDL Number MFCD00040663
InChI Key GBAOBIBJACZTNA-UHFFFAOYSA-N
Synonym calcium sulfite, calcium sulphite, sulfurous acid, calcium salt 1:1, calciumsulfit, calcium 2+ sulfite
PubChem CID 10154041
IUPAC Name calcium sulfurous acid
SMILES [Ca++].OS(O)=O

Specifications

Quantity 100 g
Formula Weight 120.14
Chemical Name or Material Calcium Sulfite