missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Silicate, NF, Spectrum™ Chemical
Description
Spectrum™ Chemical Calcium Silicate, NF , also known as calcium orthosilicate, is used as an anticaking agent in pharmaceutical preparations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Spécifications
Spécifications
| CAS Max % | 100% |
| pH | 8.4-11.2 |
| Quantity | 15 kg |
| Grade | NF |
| Loss on Ignition | 20% |
| Packaging | Fiber Drum |