missing translation for 'onlineSavingsMsg'
Learn More

Calcium Silicate, NF, Spectrum™ Chemical

Catalog No. 18601377
Change view
Click to view available options
Quantity:
100 g
1 kg
8 kg
15 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601377 100 g Poly Bottle
18601376 1 kg Poly Pail
18601379 8 kg Fiber Drum
18601380 15 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18601377 Supplier Spectrum Chemical Mfg Cor Supplier No. C1643100GM
Only null left

C1643, 1344-95-2, CaO·SiO2

Spectrum™ Chemical Calcium Silicate, NF , also known as calcium orthosilicate, is used as an anticaking agent in pharmaceutical preparations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1344-95-2
Molecular Formula CaO3Si
Molecular Weight (g/mol) 116.16
InChI Key OYACROKNLOSFPA-UHFFFAOYSA-N
IUPAC Name calcium oxosilanebis(olate)
SMILES [Ca++].[O-][Si]([O-])=O

Specifications

CAS Max % 100%
pH 8.4-11.2
Quantity 100 g
Grade NF
Loss on Ignition 20%
Packaging Poly Bottle