missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Phosphate Tribasic, Powder, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Physical Form | Crystalline Powder or Needles |
| Quantity | 45 kg |
| Grade | Reagent |
| Packaging | Poly Bottle |
| Chemical Name or Material | Calcium Phosphate Tribasic, Powder |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.