missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Phosphate Tribasic, Powder, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor C1155500GM
Specifications
| 1306-06-5 | |
| 500 g | |
| MFCD00010904 | |
| [OH-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O | |
| 502.31 | |
| Poly Bottle |
| 1 | |
| Ca5HO13P3 | |
| XYJRXVWERLGGKC-UHFFFAOYSA-D | |
| pentacalcium hydroxide triphosphate | |
| Reagent |
Chemical Identifiers
| 1306-06-5 | |
| 502.31 | |
| XYJRXVWERLGGKC-UHFFFAOYSA-D | |
| [OH-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O |
| Ca5HO13P3 | |
| MFCD00010904 | |
| pentacalcium hydroxide triphosphate |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.