missing translation for 'onlineSavingsMsg'
Learn More

Calcium phosphate (pyro), 96% min

Catalog No. p-7080527
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8983630 250 g
AA89836A1 1 kg
AA89836A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA8983630 Supplier Thermo Scientific Chemicals Supplier No. 08983630
Only null left

CAS: 7790-76-3 | Ca2O7P2 | 254.10 g/mol

Calcium phosphate is used in the production of phosphoric acid and fertilizers. It is used in baking as the acid in a leavening agent. It is also used in cheese products. This compound has also been used in gene transfection of cells. It is also being used in the development of contrast agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7790-76-3
Molecular Formula Ca2O7P2
Molecular Weight (g/mol) 254.10
MDL Number MFCD00015983
InChI Key JUNWLZAGQLJVLR-UHFFFAOYSA-J
Synonym calcium pyrophosphate, calcium diphosphate, dicalcium diphosphate, unii-x69nu20d19, dicalcium pyrophosphate, alpha-calcium pyrophosphate, calcium phosphate ca2p2o7, diphosphoric acid, calcium salt, calcium pyrophosphate ca2p2o7, diphosphoric acid, calcium salt 1:2
PubChem CID 24632
ChEBI CHEBI:32598
IUPAC Name dicalcium;phosphonato phosphate
SMILES [Ca++].[Ca++].[O-]P([O-])(=O)OP([O-])([O-])=O

Specifications

Melting Point 1230°C
Color White
Physical Form Powder
Quantity 250 g
Solubility Information Soluble in hydrochloric acid and nitric acid. Insoluble in water.
Formula Weight 254.1
Percent Purity ≥96%
Odor Odorless
Density 3.09 g/mL
Chemical Name or Material Calcium phosphate (pyro)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 232-221-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only