missing translation for 'onlineSavingsMsg'
Learn More

Calcium Phosphate, Powder, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. 02002697
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02002697 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02002697 Supplier Avantor Supplier No. 143005

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 7757-93-9
Physical Form Powder
Quantity 2.5 kg
Molecular Formula CaHO4P
MDL Number MFCD00010909
Synonym calcium hydrogen phosphate, calcium hydrogenphosphate, calcium phosphate dibasic, calcium phosphate, dibasic, biofos, fujicalin s, dicafos an, dibasic calcium phosphate, monocalcium acid phosphate, phosphoric acid, calcium salt 1:1
InChI Key FUFJGUQYACFECW-UHFFFAOYSA-L
SMILES [Ca++].OP([O-])([O-])=O
IUPAC Name calcium;hydrogen phosphate
Molecular Weight (g/mol) 136.06
PubChem CID 24441
ChEBI CHEBI:32596
Formula Weight 136.06
Grade BAKER ANALYZED™ Reagent
Packaging Poly Bottle
Specific Gravity 1L = 2.30kg
Chemical Name or Material Calcium Phosphate
Show More Show Less