missing translation for 'onlineSavingsMsg'
Learn More

Calcium Phosphate, Monobasic, Monohydrate, Crystal, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. 2002541
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
2002541 500 g Poly Bottle
02002540 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2002541 Supplier Avantor Supplier No. 142601

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 7758-23-8
Molecular Formula CaH4O8P2
Molecular Weight (g/mol) 234.05
InChI Key YYRMJZQKEFZXMX-UHFFFAOYSA-L
Synonym calcium biphosphate, monobasic calcium phosphate, calcium dihydrogen phosphate, acid calcium phosphate, primary calcium phosphate, monocalcium orthophosphate, c 38 phosphate, calcium diorthophosphate, monocalcium phosphate, monobasic, calcium dihydrogen orthophosphate
PubChem CID 24454
ChEBI CHEBI:35433
IUPAC Name calcium;dihydrogen phosphate
SMILES OP(=O)(O)[O-].OP(=O)(O)[O-].[Ca+2]

Specifications

Physical Form Crystals
Quantity 500 g
Formula Weight 252.07
Grade BAKER ANALYZED™ Reagent
Packaging Poly Bottle
Specific Gravity 1L = 2.22kg
Chemical Name or Material Calcium Phosphate, Monobasic, Monohydrate