missing translation for 'onlineSavingsMsg'
Learn More

Calcium Phosphate, Monobasic, Monohydrate, Crystal, 15.9-17.7%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor C114525KG

 View more versions of this product

Catalog No. 18609521


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

10031-30-8
2.5 kg
Reagent
Poly Bottle
1
15.9 to 17.7%
0.01

Chemical Identifiers

10031-30-8
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.