missing translation for 'onlineSavingsMsg'
Learn More

Calcium Phosphate, Monobasic, Monohydrate, Crystal, 15.9-17.7%, Spectrum™ Chemical

Catalog No. 18609519
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609519 125 g Poly Bottle
18609520 500 g Poly Bottle
18609521 2.5 kg Poly Bottle
18609522 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18609519 Supplier Spectrum Chemical Mfg Cor Supplier No. C1145125GM
Only null left

Chemical Identifiers

CAS 10031-30-8

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 15.9 to 17.7%
Grade Reagent
Loss on Drying 0.01
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.