missing translation for 'onlineSavingsMsg'
Learn More

Calcium Phosphate Dibasic, Anhydrous, Powder, 98%, Spectrum™ Chemical

Catalog No. p-7165296
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
100 lb.
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18609514 125 g Poly Bottle
18609515 500 g Poly Bottle
18609518 100 lb. Fiber Drum
18609516 2.5 kg Poly Pail
18609517 12 kg Poly Pail
5 options

Catalog No. 18609514

Supplier: Spectrum Chemical Mfg Cor C1150125GM

Only null left

Chemical Identifiers

CAS 7757-93-9

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 98%
Grade Reagent
Packaging Poly Bottle

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.