missing translation for 'onlineSavingsMsg'
Learn More

Calcium phosphate, dibasic, 98+%, for analysis, anhydrous

Catalog No. AC446432500
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC446432500 250 g Plastic bottle
AC446430500 50 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC446432500 Supplier Thermo Scientific Chemicals Supplier No. 446432500
Only null left

CAS: 7757-93-9 | CaHO4P | 136.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer

Chemical Identifiers

CAS 7757-93-9
Molecular Formula CaHO4P
Molecular Weight (g/mol) 136.06
MDL Number MFCD00010909
InChI Key FUFJGUQYACFECW-UHFFFAOYSA-L
Synonym calcium hydrogen phosphate, calcium hydrogenphosphate, calcium phosphate dibasic, calcium phosphate, dibasic, biofos, fujicalin s, dicafos an, dibasic calcium phosphate, monocalcium acid phosphate, phosphoric acid, calcium salt 1:1
PubChem CID 24441
ChEBI CHEBI:32596
IUPAC Name calcium;hydrogen phosphate
SMILES [Ca++].OP([O-])([O-])=O

Specifications

Quantity 250 g
Assay Percent Range 98.0 to 103% (Complexometry)
Linear Formula CaHPO4
Merck Index 15,1694
Solubility Information Solubility in water: 0.1g/L (25°C). Other solubilities: practically insoluble in alcohol
Formula Weight 136.06
Percent Purity ≥98%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Calcium phosphate, dibasic
EINECSNumber 231-826-1

RUO – Research Use Only