missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium phosphate, 96%, pure, mixture of Calcium phosphates
CAS: 7758-87-4 | Ca3O8P2 | 310.18 g/mol
$264.99 - $264.99
Chemical Identifiers
| CAS | 7758-87-4 |
|---|---|
| Molecular Formula | Ca3O8P2 |
| Molecular Weight (g/mol) | 310.18 |
| InChI Key | QORWJWZARLRLPR-UHFFFAOYSA-H |
| Synonym | calcium phosphate, synthos, tricalcium phosphate, tricalcium diphosphate, calcigenol simple, calcium orthophosphate, beta-tcp, tribasic calcium phosphate, bonarka, calcium phosphate, tribasic |
| PubChem CID | 24456 |
| ChEBI | CHEBI:9679 |
| IUPAC Name | tricalcium;diphosphate |
| SMILES | [O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Ca+2].[Ca+2].[Ca+2] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC446390010
|
Thermo Scientific Chemicals
446390010 |
1 kg |
Each for $264.99
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7758-87-4 | |
| 310.18 | |
| calcium phosphate, synthos, tricalcium phosphate, tricalcium diphosphate, calcigenol simple, calcium orthophosphate, beta-tcp, tribasic calcium phosphate, bonarka, calcium phosphate, tribasic | |
| CHEBI:9679 | |
| [O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Ca+2].[Ca+2].[Ca+2] |
| Ca3O8P2 | |
| QORWJWZARLRLPR-UHFFFAOYSA-H | |
| 24456 | |
| tricalcium;diphosphate |
Specifications
| 1670.0°C | |
| 1 kg | |
| Ca3O8P2 | |
| 15,1696 | |
| Solubility in water: 0.1g/L (25°C) | |
| [O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Ca+2].[Ca+2].[Ca+2] | |
| 310.18 | |
| CHEBI:9679 | |
| 96% | |
| Glass bottle |
| 7758-87-4 | |
| 95% min. (Complexometry) | |
| Ca3(PO4)2 | |
| calcium phosphate, synthos, tricalcium phosphate, tricalcium diphosphate, calcigenol simple, calcium orthophosphate, beta-tcp, tribasic calcium phosphate, bonarka, calcium phosphate, tribasic | |
| QORWJWZARLRLPR-UHFFFAOYSA-H | |
| tricalcium;diphosphate | |
| 24456 | |
| 310.18 | |
| Pure | |
| Calcium phosphate |
Safety and Handling
EINECSNumber : 231-840-8
RUO – Research Use Only