missing translation for 'onlineSavingsMsg'
Learn More

Calcium Pantothenate, FCC, 97-103%, Spectrum™ Chemical

Catalog No. 18601333
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Amber Glass Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601333 100 g Amber Glass Bottle
18601334 500 g Amber Glass Bottle
18601335 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18601333 Supplier Spectrum Chemical Mfg Cor Supplier No. C1143100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

C1143, 137-08-6, C18H32CaN2O10

Spectrum™ Chemical Calcium Pantothenate, FCC is often used in dietary supplements because, as a salt, it is more stable than pantothenic acid (Vitamin B5). The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 137-08-6
Molecular Formula C18H32CaN2O10
Molecular Weight (g/mol) 476.54
MDL Number MFCD00002766
InChI Key FAPWYRCQGJNNSJ-DXHDTSSINA-L
IUPAC Name calcium bis(3-[(2R)-2,4-dihydroxy-3,3-dimethylbutanamido]propanoate)
SMILES [Ca++].CC(C)(CO)[C@@H](O)C(=O)NCCC([O-])=O.CC(C)(CO)[C@@H](O)C(=O)NCCC([O-])=O

Specifications

CAS Max % 100%
Loss on Drying 5%
Packaging Amber Glass Bottle
Specific Rotation +25.0° to +27.5°
Quantity 100 g
Percent Purity 97 to 103%
Grade FCC