missing translation for 'onlineSavingsMsg'
Learn More

Calcium Oxide, Purified (Lime), Reagents

Catalog No. p-200021146
Change view
Click to view available options
Quantity:
500 g
3 kg
12 kg
Catalog No. Quantity
01337331 12 kg
01337330 3 kg
01337329 500 g
3 options

Catalog No. 01337331

Supplier: Labchem Inc C216600012A5

Only null left

Calcium Oxide, Purified (Lime), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1305-78-8
Molecular Formula CaO
Molecular Weight (g/mol) 56.08
MDL Number MFCD00010911
InChI Key BRPQOXSCLDDYGP-UHFFFAOYSA-N
Synonym Lime, Quicklime, Burnt lime
IUPAC Name calcium oxidandiide
SMILES [O--].[Ca++]

Specifications

Boiling Point 2850°C
Melting Point 2570°C
Color White to Off-White
Physical Form Solid
Quantity 12 kg
Grade Pure
Packaging Poly Pail
Specific Gravity 3.34
Chemical Name or Material Calcium Oxide, Purified (Lime)