missing translation for 'onlineSavingsMsg'
Learn More

Calcium oxide, Puratronic™, 99.998% (metals basis, excluding other alkaline earth and alkali metals 130ppm max)

Catalog No. p-3256944
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
250 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1068404 2 g
AA1068409 10 g
AA1068418 50 g
AA1068430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA1068404 Supplier Thermo Scientific Chemicals Supplier No. 01068404
Only null left

CAS: 1305-78-8 | CaO | 56.08 g/mol

Calcium oxide has wide-ranging utility in industry. It is used in making porcelain, cements, mortars, and glass. It is also employed in flue-gas desulfurization in power plants, purifying sugar, drying the solvents/reagents, preparing bleaching powder, calcium carbide, and calcium cyanamide. It also finds use in water softeners, chemical recovery at Kraft pulp mills, biodiesel production, and in petroleum industry for the detection of traces of water. In agriculture it is used to restore the pH of acidic soils (liming). It is useful as a basic catalyst & acid scavenger in several organic transformations, for example, in Arndt-Eistert Synthesis of alpha-diazoketones, synthesis of heterocycles and oximes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1305-78-8
Molecular Formula CaO
Molecular Weight (g/mol) 56.08
MDL Number MFCD00010911
InChI Key BRPQOXSCLDDYGP-UHFFFAOYSA-N
Synonym calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, airlock, calx
PubChem CID 14778
ChEBI CHEBI:31344
SMILES [O--].[Ca++]

Specifications

Boiling Point 2850°C
Melting Point 2572°C
Color White
Physical Form Powder
Quantity 2 g
Assay Percent Range 99.998% (Metal basis)
Solubility Information Slightly soluble in water. Insoluble in ethanol. Soluble in glycerol.
IUPAC Name oxocalcium
Formula Weight 56.08
Odor Odorless
Refractive Index 1.83
Sensitivity Air and moisture sensitive
Appearance White powder
Density 3.37
Chemical Name or Material Calcium oxide
Show More Show Less
Hazard Category H315-H318-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
DOTInformation DOT Class: 8, Packaging Group: III
EINECSNumber 215-138-9
RTECSNumber EW3100000
TSCA Yes
storageNote1 Air & Moisture Sensitive
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only