missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Oxide, Honeywell Fluka™
Puriss., meets analytical specification of FCC, 96-100.5% (ex ignited substance), powder (fine)
Supplier: Honeywell Chemicals 120475KG
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Calcium Oxide | |
| 1305-78-8 | |
| 5 kg | |
| CaO | |
| 1910 | |
| BRPQOXSCLDDYGP-UHFFFAOYSA-N | |
| oxocalcium | |
| 14778 | |
| 56.08g/mol | |
| puriss. |
| Meets analytical specification of FCC | |
| Fine Powder | |
| CaO | |
| MFCD00010911 | |
| calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, airlock, calx | |
| [O--].[Ca++] | |
| 56.08 | |
| CHEBI:31344 | |
| 96 to 100.5% (ex ignited substance) | |
| 3.3g/mL at 25°C |
Chemical Identifiers
| 1305-78-8 | |
| 56.08 | |
| BRPQOXSCLDDYGP-UHFFFAOYSA-N | |
| 14778 | |
| [O--].[Ca++] |
| CaO | |
| MFCD00010911 | |
| calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, airlock, calx | |
| CHEBI:31344 |
Safety and Handling
P261-P280-P305 + P351 + P338
H315-H318-H335
EINECSNumber : 215-138-9
RTECSNumber : EW3100000