missing translation for 'onlineSavingsMsg'
Learn More

Calcium Nitrate Tetrahydrate (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. C1093
Change view
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
C1093 3 kg Poly Bottle
C109500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1093 Supplier Thermo Fisher Scientific Supplier No. FLC1093
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13477-34-4
Molecular Formula CaH8N2O10
Molecular Weight (g/mol) 236.15
MDL Number MFCD00149604
InChI Key MWGCGFYACDTFSB-UHFFFAOYSA-N
Synonym calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate
PubChem CID 16211656
ChEBI CHEBI:86159
IUPAC Name calcium bis(nitric acid) tetrahydrate
SMILES O.[Ca++].[O-][N+]([O-])=O

Specifications

Boiling Point 132°C
Melting Point 45°C
Color White
pH 5 to 7
Physical Form Solid
Quantity 3 kg
Assay Percent Range 99 to 103 %
Linear Formula Ca(NO3)2·4H2O
Formula Weight 236.15
Percent Purity 99 to 103%
Grade Certified ACS
Identification Pass Test
Packaging Poly Bottle
Density 1.860g/cm³
Chemical Name or Material Calcium Nitrate Tetrahydrate
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 5.1, : Oxidizer