missing translation for 'onlineSavingsMsg'
Learn More

Calcium Nitrate, Tetrahydrate, Crystal, ACS, 99-103%, Spectrum™ Chemical

Catalog No. 18609500
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609500 25 g Poly Bottle
18609501 125 g Poly Bottle
18609502 500 g Poly Bottle
18609503 2.5 kg Poly Pail
18609504 12 kg Poly Pail
18609505 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18609500 Supplier Spectrum Chemical Mfg Cor Supplier No. C113525GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 13477-34-4
Molecular Formula CaH8N2O10
Molecular Weight (g/mol) 236.15
InChI Key MWGCGFYACDTFSB-UHFFFAOYSA-N
IUPAC Name calcium bis(nitric acid) tetrahydrate
SMILES O.[Ca++].[O-][N+]([O-])=O

Specifications

CAS Max % 1
pH 5.0 to 7.0
Quantity 25 g
Percent Purity 99 to 103%
Grade ACS
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.