missing translation for 'onlineSavingsMsg'
Learn More

Calcium Nitrate Tetrahydrate, ACS Reagent, Reagents

Numéro de catalogue. 01338060
Change view
Click to view available options
Quantity:
125 g
50 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
01338060 125 g
01338058 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 01338060 Fournisseur Labchem Inc Code fournisseur C1165500125D2
Il en reste null

Calcium Nitrate Tetrahydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Identifiants chimiques

CAS 13477-34-4
Molecular Formula CaH8N2O10
Molecular Weight (g/mol) 236.15
InChI Key MWGCGFYACDTFSB-UHFFFAOYSA-N
Synonym Nitric Acid calcium salt
IUPAC Name calcium bis(nitric acid) tetrahydrate
SMILES O.[Ca++].[O-][N+]([O-])=O

Spécifications

Boiling Point 132°C
Melting Point 561°C
Color Colorless/White
Physical Form Solid
Quantity 125 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 1.82
Chemical Name or Material Calcium Nitrate Tetrahydrate, ACS Reagent