missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium nitrate tetrahydrate, ACS, 99.0-103.0%
Description
Calcium nitrate tetrahydrate is used as nitrogenous fertilizer and corrosive inhibitor in diesel fuels. It is utilized in plant cell for culture experiments. It is also used in cooling baths and waste water handling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 132°C (decomposition) |
| Melting Point | 42°C |
| Physical Form | Crystalline |
| Quantity | 500 g |
| Assay Percent Range | 99.0-103.0% |
| UN Number | UN1454 |
| Merck Index | 14,1682 |
| Solubility Information | Soluble in water,alcohol,acetone and ammonia. |
| Formula Weight | 236.15 (164.09 Anhydrous) |
| Percent Purity | 99.0 to 103.0% |
| Show More |
Safety and Handling
| Hazard Category | H302-H318 |
| Hazard Statement | GHS H Statement H272-H315-H319-H335 May intensify fire oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c |
| DOTInformation | Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: CALCIUM NITRATE |
| EINECSNumber | 233-332-1 |
| RTECSNumber | EW3000000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only