missing translation for 'onlineSavingsMsg'
Learn More

Calcium Nitrate Tetrahydrate, ≥99%, (ACS Reagent), MP Biomedicals

Supplier:  MP Biomedicals Inc 0219380180

Catalog No. ICN19380180

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Calcium Nitrate Tetrahydrate
100 g
CaH8N2O10
MWGCGFYACDTFSB-UHFFFAOYSA-N
calcium bis(nitric acid) tetrahydrate
16211656
236.2
13477-34-4
≥99%
calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate
O.[Ca++].[O-][N+]([O-])=O
236.15
CHEBI:86159
ACS Reagent

Chemical Identifiers

13477-34-4
236.15
calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate
CHEBI:86159
O.[Ca++].[O-][N+]([O-])=O
CaH8N2O10
MWGCGFYACDTFSB-UHFFFAOYSA-N
16211656
calcium bis(nitric acid) tetrahydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions