missing translation for 'onlineSavingsMsg'
Learn More

Calcium nitrate tetrahydrate, 99.98% (metals basis)

Catalog No. p-4407336
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3048214 25 g
AA3048222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3048214 Supplier Thermo Scientific Chemicals Supplier No. 03048214
Only null left

CAS: 13477-34-4 | CaH8N2O10 | 236.15 g/mol

Calcium nitrate tetrahydrate is used as a nitrogenous fertilizer and a corrosive inhibitor in diesel fuels. It is utilized in plant cells for culture experiments. It is also used in cooling baths and waste water handling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13477-34-4
Molecular Formula CaH8N2O10
Molecular Weight (g/mol) 236.15
MDL Number MFCD00149604
InChI Key MWGCGFYACDTFSB-UHFFFAOYSA-N
Synonym calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate
PubChem CID 16211656
ChEBI CHEBI:86159
IUPAC Name calcium bis(nitric acid) tetrahydrate
SMILES O.[Ca++].[O-][N+]([O-])=O

Specifications

Boiling Point 132°C (decomposition)
Melting Point 42°C
Color White
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99.98%
Solubility Information Soluble in water,alcohol and acetone.
Formula Weight 236.15 (164.09 Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Appearance White crystalline or powder
Density 1.82 g/cm3 at 20°C
Chemical Name or Material Calcium nitrate tetrahydrate
Show More Show Less
Hazard Category H302-H318
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
DOTInformation DOT Class: 5.1, Packaging Group: III
EINECSNumber 233-332-1
RTECSNumber EW3000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only