Learn More
Calcium nitrate tetrahydrate, 98%, extra pure
CAS: 13477-34-4 | CaH8N2O10 | 236.15 g/mol
Supplier: Thermo Scientific Chemicals 447530050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 132.0°C | |
| 45.0°C | |
| 5.0 to 9.0 | |
| 97.5% min. (Complexometry) | |
| Ca(NO3)2·4H2O | |
| calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate | |
| MWGCGFYACDTFSB-UHFFFAOYSA-N | |
| calcium bis(nitric acid) tetrahydrate | |
| 16211656 | |
| 236.15 | |
| Extra Pure | |
| 1.8600g/mL |
| Calcium nitrate tetrahydrate | |
| 13477-34-4 | |
| 5 kg | |
| CaH8N2O10 | |
| 15,1684 | |
| Solubility in water: 1470g/L (0°C). Other solubilities: soluble in alcohol and acetone | |
| O.[Ca++].[O-][N+]([O-])=O | |
| 236.15 | |
| CHEBI:86159 | |
| 98% | |
| 1.86 |
Chemical Identifiers
| 13477-34-4 | |
| 236.15 | |
| calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate | |
| CHEBI:86159 | |
| O.[Ca++].[O-][N+]([O-])=O |
| CaH8N2O10 | |
| MWGCGFYACDTFSB-UHFFFAOYSA-N | |
| 16211656 | |
| calcium bis(nitric acid) tetrahydrate |
Safety and Handling
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
RUO – Research Use Only