Learn More
Calcium nitrate tetrahydrate, 97+%
CAS: 13477-34-4 | CaH8N2O10 | 236.15 g/mol
Supplier: Thermo Scientific Chemicals A1664536
Description
Calcium nitrate tetrahydrate is used as a nitrogenous fertilizer and a corrosive inhibitor in diesel fuels. It is utilized in plant cells for culture experiments. It is also used in cooling baths and waste water handling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCalcium nitrate tetrahydrate is used as a nitrogenous fertilizer and a corrosive inhibitor in diesel fuels. It is utilized in plant cells for culture experiments. It is also used in cooling baths and waste water handling.
Solubility
Soluble in water, alcohol and acetone.
Notes
Store in cool place. Hygroscopic. Incompatible with strong reducing agents.
Specifications
| 132°C (decomposition) | |
| 43°C to 45°C | |
| 500 g | |
| MFCD00149604 | |
| 14,1682 | |
| Soluble in water,alcohol and acetone. | |
| O.[Ca++].[O-][N+]([O-])=O | |
| 236.15 | |
| CHEBI:86159 | |
| 99% | |
| Hygroscopic |
| Calcium nitrate tetrahydrate | |
| 13477-34-4 | |
| CaH8N2O10 | |
| UN1454 | |
| calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate | |
| MWGCGFYACDTFSB-UHFFFAOYSA-N | |
| calcium bis(nitric acid) tetrahydrate | |
| 16211656 | |
| 236.15 (164.09 Anhydrous) | |
| Odorless | |
| 1.82 |
Chemical Identifiers
| 13477-34-4 | |
| 236.15 | |
| MWGCGFYACDTFSB-UHFFFAOYSA-N | |
| 16211656 | |
| calcium bis(nitric acid) tetrahydrate |
| CaH8N2O10 | |
| MFCD00149604 | |
| calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate | |
| CHEBI:86159 | |
| O.[Ca++].[O-][N+]([O-])=O |
Safety and Handling
GHS H Statement
H272-H315-H319-H335
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
H302-H318
DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: CALCIUM NITRATE
EINECSNumber : 233-332-1
RTECSNumber : EW3000000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only