Learn More
Calcium nitrate hydrate, Puratronic™, 99.9995% (metals basis)
CAS: 35054-52-5 | CaN2O6 | 164.09 g/mol
$96.04 - $979.74
Chemical Identifiers
| CAS | 35054-52-5 |
|---|---|
| Molecular Formula | CaN2O6 |
| Molecular Weight (g/mol) | 164.09 |
| MDL Number | MFCD00149604 |
| InChI Key | ZCCIPPOKBCJFDN-UHFFFAOYSA-N |
| Synonym | calcium nitrate hydrate, calcium hydrate dinitrate, acmc-20alid, calcium nitrate monohydrate, ca.2no3.h2o, nitric acid calcium salt hydrate, calcium nitrate-water 1/2/1, nitric acid, calciumsalt, hydrate 9ci |
| PubChem CID | 16211464 |
| SMILES | [Ca++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA4451506
|
Thermo Scientific Chemicals
04451506 |
5 g |
Each for $96.04
|
|
|||||
|
AA4451514
|
Thermo Scientific Chemicals
04451514 |
25 g |
Each for $330.11
|
|
|||||
|
AA4451522
|
Thermo Scientific Chemicals
04451522 |
100 g |
Each for $979.74
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 35054-52-5 | |
| 164.09 | |
| ZCCIPPOKBCJFDN-UHFFFAOYSA-N | |
| 16211464 |
| CaN2O6 | |
| MFCD00149604 | |
| calcium nitrate hydrate, calcium hydrate dinitrate, acmc-20alid, calcium nitrate monohydrate, ca.2no3.h2o, nitric acid calcium salt hydrate, calcium nitrate-water 1/2/1, nitric acid, calciumsalt, hydrate 9ci | |
| [Ca++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
| 132°C (decomposition) | |
| 35054-52-5 | |
| Crystalline | |
| CaN2O6 | |
| MFCD00149604 | |
| 14,1682 | |
| ZCCIPPOKBCJFDN-UHFFFAOYSA-N | |
| calcium dinitrate | |
| 16211464 | |
| 99.9995% | |
| 1.82 g/mL | |
| (metals basis) |
| 42°C | |
| White | |
| 5 g | |
| Ca(NO3)2·xH2O | |
| UN1454 | |
| calcium nitrate hydrate, calcium hydrate dinitrate, acmc-20alid, calcium nitrate monohydrate, ca.2no3.h2o, nitric acid calcium salt hydrate, calcium nitrate-water 1/2/1, nitric acid, calciumsalt, hydrate 9ci | |
| [Ca++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
| 164.09 | |
| 164.09 (Anhydrous) | |
| Puratronic | |
| Calcium nitrate hydrate |
Safety and Handling
GHS H Statement
H272-H302-H315-H319-H335
May intensify fire
oxidizer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P220-P221-P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P370+P378q-P501c
H272-H302-H318
DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: CALCIUM NITRATE
EINECSNumber : 233-332-1
RTECSNumber : EW3000000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only