Learn More
Calcium molybdenum oxide, 99%
CAS: 7789-82-4 | CaMoO4 | 200.02 g/mol
Supplier: Thermo Scientific Chemicals 04368614
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Calcium molybdenum oxide | |
| 7789-82-4 | |
| Powder | |
| CaMoO4 | |
| 14,1681 | |
| BIOOACNPATUQFW-UHFFFAOYSA-N | |
| calcium;dioxido(dioxo)molybdenum | |
| 24620 | |
| 99% | |
| 4.45 g/mL |
| 965°C | |
| White | |
| 25 g | |
| MFCD00015976 | |
| calcium molybdate, molybdate, calcium, calcium molybdate vi, unii-q03c83v2an, calcium molybdenum oxide, molybdic acid h2moo4 , calcium salt 1:1, molybdate moo42-, calcium 1:1 , t-4, molybdate moo42-, calcium 1:1 , beta-4, calcium dioxido dioxo molybdenum, calcium 2+ molybdate | |
| [Ca++].[O-][Mo]([O-])(=O)=O | |
| 200.02 | |
| 200.01 | |
| Odorless |
Chemical Identifiers
| 7789-82-4 | |
| 200.02 | |
| BIOOACNPATUQFW-UHFFFAOYSA-N | |
| 24620 |
| CaMoO4 | |
| MFCD00015976 | |
| calcium molybdate, molybdate, calcium, calcium molybdate vi, unii-q03c83v2an, calcium molybdenum oxide, molybdic acid h2moo4 , calcium salt 1:1, molybdate moo42-, calcium 1:1 , t-4, molybdate moo42-, calcium 1:1 , beta-4, calcium dioxido dioxo molybdenum, calcium 2+ molybdate | |
| [Ca++].[O-][Mo]([O-])(=O)=O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 232-192-9
RTECSNumber : EW2975000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only