missing translation for 'onlineSavingsMsg'
Learn More

Calcium Methanesulfonate 98.0+%, TCI America™

Catalog No. M0549500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0549500G 500 g
M054925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0549500G Supplier TCI America Supplier No. M0549500G

Chemical Identifiers

CAS 58131-47-8
Molecular Formula C2H6CaO6S2
Molecular Weight (g/mol) 230.262
MDL Number MFCD00070543
InChI Key BWEYVLQUNDGUEC-UHFFFAOYSA-L
Synonym Methanesulfonic Acid Calcium Salt
PubChem CID 14274928
IUPAC Name calcium;methanesulfonate
SMILES CS(=O)(=O)[O-].CS(=O)(=O)[O-].[Ca+2]

Specifications

Color White
Quantity 500 g
Formula Weight 230.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Calcium Methanesulfonate