missing translation for 'onlineSavingsMsg'
Learn More

Calcium Lactate, Pentahydrate, FCC, EP, USP, 98-101%, Spectrum™ Chemical

Catalog No. p-7157210
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601326 500 g Poly Bottle
18601327 2.5 kg Poly Pail
18601328 12 kg Poly Pail
18601329 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18601326 Supplier Spectrum Chemical Mfg Cor Supplier No. C2106500GM

C2106, 5743-47-5, C6H10CaO6·5H2O

Spectrum™ Chemical Calcium Lactate, Pentahydrate, USP, FCC, EP is most commonly used as an antacid and to treat calcium deficiencies. Spectrum Chemical manufactured USP, FCC, EP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 5743-47-5
Molecular Formula C6H20CaO11
Molecular Weight (g/mol) 308.29
MDL Number MFCD00287281 (5H2O)
InChI Key HPVJXNNKHRNBOY-UHFFFAOYNA-L
IUPAC Name calcium bis(2-hydroxypropanoate) pentahydrate
SMILES O.O.O.O.O.[Ca++].CC(O)C([O-])=O.CC(O)C([O-])=O

Specifications

CAS Max % 100%
Loss on Drying 22.0-27%
Packaging Poly Bottle
Quantity 500 g
Percent Purity 98-101%
Grade Multi Compendia