missing translation for 'onlineSavingsMsg'
Learn More

Calcium(II) Bis(trifluoromethanesulfonyl)imide 97.0+%, TCI America™

Catalog No. C32631G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C32631G 1 g
C32635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C32631G Supplier TCI America Supplier No. C32631G

Chemical Identifiers

CAS 165324-09-4
Molecular Formula C4CaF12N2O8S4
Molecular Weight (g/mol) 600.349
MDL Number MFCD14156019
InChI Key WFABOCFDABTAPE-UHFFFAOYSA-N
Synonym calcium bis trifluoromethylsulfonyl imide, calcium ii bis trifluoromethanesulfonyl imide, calcium ii triflimide, calcium trifluoromethanesulfonimide, calcium bistrifluoromethanesulfonimide, calcium bis trifluoromethylsulfonyl amide, calcium bis bistriflylimide anion, calcium bis trifluoromethyl sulfonyl amide, bis bis trifluoromethylsulfonyl amino calcium, bis trifluoromethanesulfonyl imide calcium ii salt
PubChem CID 46242197
IUPAC Name calcium;bis(trifluoromethylsulfonyl)azanide
SMILES C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Ca+2]

Specifications

Color White
Quantity 1 g
UN Number 1759
Formula Weight 600.35
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Calcium(II) Bis(trifluoromethanesulfonyl)imide