missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Hypochlorite, Reagents
Calcium Hypochlorite, Technical, for general laboratory use. Manufactured in ISO 9001 facility.
$159.04 - $361.11
Chemical Identifiers
| CAS | 7778-54-3 |
|---|---|
| Molecular Formula | CaCl2O2 |
| Molecular Weight (g/mol) | 142.98 |
| InChI Key | ZKQDCIXGCQPQNV-UHFFFAOYSA-N |
| Synonym | Bleaching powder, Chlorinated lime, Hypochlorous Acid calcium salt |
| IUPAC Name | calcium dihypochlorite |
| SMILES | [Ca++].[O-]Cl.[O-]Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01-337-333
|
Labchem Inc
C21650003A5 |
3 kg |
Each for $361.11
|
|
|||||
|
01-337-332
|
Labchem Inc
C2165000500A5 |
500 g |
Each for $159.04
|
|
|||||
Chemical Identifiers
| 7778-54-3 | |
| 142.98 | |
| Bleaching powder, Chlorinated lime, Hypochlorous Acid calcium salt | |
| [Ca++].[O-]Cl.[O-]Cl |
| CaCl2O2 | |
| ZKQDCIXGCQPQNV-UHFFFAOYSA-N | |
| calcium dihypochlorite |
Specifications
| 100°C | |
| 7778-54-3 | |
| 3 kg | |
| Bleaching powder, Chlorinated lime, Hypochlorous Acid calcium salt | |
| [Ca++].[O-]Cl.[O-]Cl | |
| 142.98 | |
| Poly Bottle | |
| Calcium Hypochlorite |
| Colorless/White | |
| Solid | |
| CaCl2O2 | |
| ZKQDCIXGCQPQNV-UHFFFAOYSA-N | |
| calcium dihypochlorite | |
| Technical | |
| 2.35 |