missing translation for 'onlineSavingsMsg'
Learn More

Calcium Hypochlorite, Reagents

Catalog No. 01337333
Change view
Click to view available options
Quantity:
500 g
3 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337333 3 kg
01337332 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337333 Supplier Labchem Inc Supplier No. C21650003A5

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Calcium Hypochlorite, Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7778-54-3
Molecular Formula CaCl2O2
Molecular Weight (g/mol) 142.98
InChI Key ZKQDCIXGCQPQNV-UHFFFAOYSA-N
Synonym Bleaching powder, Chlorinated lime, Hypochlorous Acid calcium salt
IUPAC Name calcium dihypochlorite
SMILES [Ca++].[O-]Cl.[O-]Cl

Specifications

Melting Point 100°C
Color Colorless/White
Physical Form Solid
Quantity 3 kg
Grade Technical
Packaging Poly Bottle
Specific Gravity 2.35
Chemical Name or Material Calcium Hypochlorite