missing translation for 'onlineSavingsMsg'
Learn More

Calcium Hydroxide, purum p.a., Honeywell™

Catalog No. p-7071868
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002086 1 kg
6002087 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6002086 Supplier Honeywell Chemicals Supplier No. 211811KG
Only null left

Purum p.a.,≥96.0% (T)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.

Chemical Identifiers

CAS 1305-62-0
Molecular Formula CaH2O2
Molecular Weight (g/mol) 74.09
MDL Number MFCD00010901
InChI Key AXCZMVOFGPJBDE-UHFFFAOYSA-L
Synonym calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid
PubChem CID 6093208
ChEBI CHEBI:31341
IUPAC Name calcium;dihydroxide
SMILES [OH-].[OH-].[Ca++]

Specifications

Quantity 1 kg
Linear Formula Ca(OH)2
UN Number NONH for all modes of transport
Formula Weight 74.09g/mol
Percent Purity ≥96% (T)
Grade purum p.a.
Density 2.24g/mL at 25°C
Chemical Name or Material Calcium Hydroxide
Hazard Statement P261-P280-P305 + P351 + P338
Precautionary Statement H315-H318-H335
EINECSNumber 215-137-3
RTECSNumber EW2800000