missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Hydroxide, Purified (Slaked Lime), Reagents
Specifications
Specifications
| Boiling Point | 2850°C |
| Melting Point | 580°C |
| Color | Colorless/White |
| Physical Form | Solid |
| Quantity | 2.5 kg |
| Grade | Pure |
| Packaging | Poly Pail |
| Specific Gravity | 2.24 |
| Chemical Name or Material | Calcium Hydroxide, Purified (Slaked Lime) |