missing translation for 'onlineSavingsMsg'
Learn More

Calcium Hydroxide, Powder, FCC, 95-100.5%, Spectrum™ Chemical

Catalog No. 18601316
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601316 500 g Poly Bottle
18601317 2.5 kg Poly Pail
18601318 12 kg Poly Pail
18601319 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18601316 Supplier Spectrum Chemical Mfg Cor Supplier No. C1116500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

C1116, 1305-62-0, Ca(OH)2

Spectrum™ Chemical Calcium Hydroxide, Powder, FCC is used extensively in the food industry to clarify solutions, as a digestion aid, baking soda substitute and as a nutritional supplement because of its low toxicity. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1305-62-0
Molecular Formula CaH2O2
Molecular Weight (g/mol) 74.09
MDL Number MFCD00010901
InChI Key AXCZMVOFGPJBDE-UHFFFAOYSA-L
IUPAC Name calcium dihydroxide
SMILES [OH-].[OH-].[Ca++]

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 95 to 100.5%
Grade FCC
Packaging Poly Bottle