missing translation for 'onlineSavingsMsg'
Learn More

Calcium Hydroxide, Powder, ACS, 95%, Spectrum™ Chemical

Catalog No. p-7165283
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
12 kg
2.5 kg
50 kg
500 g
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18609486 500 g Poly Bottle
18609487 2.5 kg Poly Pail
18609488 12 kg Poly Pail
18609489 50 kg Fiber Drum
4 options

Catalog No. 18609486

Supplier: Spectrum Chemical Mfg Cor C1115500GM

Only null left

Chemical Identifiers

CAS 1305-62-0
Molecular Formula CaH2O2
Molecular Weight (g/mol) 74.09
MDL Number MFCD00010901
InChI Key AXCZMVOFGPJBDE-UHFFFAOYSA-L
IUPAC Name calcium dihydroxide
SMILES [OH-].[OH-].[Ca++]

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 95%
Grade ACS
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.