missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Hydroxide, Powder, ACS, 95%, Spectrum™ Chemical
Specifications
Specifications
| Physical Form | Crystalline Powder or Needles |
| CAS Max % | 1 |
| Quantity | 50 kg |
| Packaging | Poly Bottle |
| Percent Purity | 95% |
| Grade | ACS/Reagent |
| Chemical Name or Material | Calcium Hydroxide, Powder |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.