missing translation for 'onlineSavingsMsg'
Learn More

Calcium Hydroxide, ACS Reagent, Reagents

Catalog No. 01338062
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01338062 2.5 kg
01338061 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01338062 Supplier Labchem Inc Supplier No. C11645002.5KG
Only null left

Calcium Hydroxide, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1305-62-0
Molecular Formula CaH2O2
Molecular Weight (g/mol) 74.09
MDL Number MFCD00010901
InChI Key AXCZMVOFGPJBDE-UHFFFAOYSA-L
Synonym Hydrated Lime
IUPAC Name calcium dihydroxide
SMILES [OH-].[OH-].[Ca++]

Specifications

Boiling Point 2850°C
Melting Point 580°C
Color Colorless/White
Physical Form Solid
Quantity 2.5 kg
Percent Purity Min. 95%
Grade ACS
Packaging Poly Pail
Specific Gravity 2.24
Chemical Name or Material Calcium Hydroxide, ACS Reagent