missing translation for 'onlineSavingsMsg'
Learn More

Calcium Hydroxide, ACS Reagent, Reagents

Catalog No. 01338061
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01338061 500 g
01338063 12 kg
01338062 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01338061 Supplier Labchem Inc Supplier No. C1164500500A5

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Calcium Hydroxide, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1305-62-0
Molecular Formula CaH2O2
Molecular Weight (g/mol) 74.09
MDL Number MFCD00010901
InChI Key AXCZMVOFGPJBDE-UHFFFAOYSA-L
Synonym Hydrated Lime
IUPAC Name calcium dihydroxide
SMILES [OH-].[OH-].[Ca++]

Specifications

Boiling Point 2850°C
Melting Point 580°C
Color Colorless/White
Physical Form Solid
Quantity 500 g
Percent Purity Min. 95%
Grade ACS
Packaging Poly Bottle
Specific Gravity 2.24
Chemical Name or Material Calcium Hydroxide, ACS Reagent