missing translation for 'onlineSavingsMsg'
Learn More

Calcium hydroxide, 95%

CAS: 1305-62-0 | CaH2O2 | 74.09 g/mol

Supplier:  Thermo Scientific Chemicals A1265030

 View more versions of this product

Catalog No. AAA1265030


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Calcium hydroxide is used as a flocculant in water and sewage treatment. It is also used in fresh water treatment to raise the pH of the water thereby controlling the corrosion of pipes. It is widely used in the production of calcium stearate. It is an active ingredient in whitewash, mortar and plaster. This compound is also used in the petroleum refining industry for the preparation of additives to oils and in the preparation of dry mixes for painting and decorating. It finds application as an antifungal and antimicrobial preservative for vegetables in storage.

Solubility
Soluble in glycerol and acids. Slightly soluble in water.Insoluble in alcohol.

Notes
Air sensitive. Incompatible with strong acids.Incompatible with maleic anhydride, phosphorous, nitroethane, nitromethane, nitorparaffins, nitropropane and strong acids.
Specifications

Specifications

Calcium hydroxide
250 g
Ca(OH)2
UN3262
calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid
AXCZMVOFGPJBDE-UHFFFAOYSA-L
calcium;dihydroxide
6093208
74.09
Odorless
2.24
1305-62-0
CaH2O2
MFCD00010901
14,1673
Soluble in glycerol and acids. Slightly soluble in water.Insoluble in alcohol.
[OH-].[OH-].[Ca++]
74.09
CHEBI:31341
95%
Air Sensitive

Chemical Identifiers

1305-62-0
74.09
AXCZMVOFGPJBDE-UHFFFAOYSA-L
6093208
calcium;dihydroxide
CaH2O2
MFCD00010901
calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid
CHEBI:31341
[OH-].[OH-].[Ca++]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, BASIC, INORGANIC, N.O.S.

EINECSNumber : 215-137-3

RTECSNumber : EW2800000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only