missing translation for 'onlineSavingsMsg'
Learn More

Calcium hydroxide, 95%

Catalog No. AAA126500B
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA126500B 1000 g
AAA1265030 250 g
AAA126500I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA126500B Supplier Thermo Scientific Chemicals Supplier No. A126500B

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1305-62-0 | CaH2O2 | 74.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Calcium hydroxide is used as a flocculant in water and sewage treatment. It is also used in fresh water treatment to raise the pH of the water thereby controlling the corrosion of pipes. It is widely used in the production of calcium stearate. It is an active ingredient in whitewash, mortar and plaster. This compound is also used in the petroleum refining industry for the preparation of additives to oils and in the preparation of dry mixes for painting and decorating. It finds application as an antifungal and antimicrobial preservative for vegetables in storage.

Solubility
Soluble in glycerol and acids. Slightly soluble in water.Insoluble in alcohol.

Notes
Air sensitive. Incompatible with strong acids.Incompatible with maleic anhydride, phosphorous, nitroethane, nitromethane, nitorparaffins, nitropropane and strong acids.

Chemical Identifiers

CAS 1305-62-0
Molecular Formula CaH2O2
Molecular Weight (g/mol) 74.09
MDL Number MFCD00010901
InChI Key AXCZMVOFGPJBDE-UHFFFAOYSA-L
Synonym calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid
PubChem CID 6093208
ChEBI CHEBI:31341
IUPAC Name calcium;dihydroxide
SMILES [OH-].[OH-].[Ca++]

Specifications

Quantity 1000 g
Odor Odorless
Sensitivity Air Sensitive
Density 2.24
Linear Formula Ca(OH)2
UN Number UN3262
Merck Index 14,1673
Solubility Information Soluble in glycerol and acids. Slightly soluble in water.Insoluble in alcohol.
Formula Weight 74.09
Percent Purity 95%
Chemical Name or Material Calcium hydroxide
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, BASIC, INORGANIC, N.O.S.
EINECSNumber 215-137-3
RTECSNumber EW2800000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only