missing translation for 'onlineSavingsMsg'
Learn More

Calcium hydrogen phosphate dihydrate, 98% min

CAS: 7789-77-7 | CaH5O6P | 172.086 g/mol

$164.70 - $501.34

Chemical Identifiers

CAS 7789-77-7
Molecular Formula CaH5O6P
Molecular Weight (g/mol) 172.086
MDL Number MFCD00149621
InChI Key XAAHAAMILDNBPS-UHFFFAOYSA-L
Synonym calcium hydrogenphosphate dihydrate, calcium hydrogen phosphate dihydrate, unii-o7tsz97gep, calcium monohydrogen phosphate dihydrate, o7tsz97gep, calcium phosphate dibasic dihydrate, dibasic calcium phosphate dihydrate, phosphoric acid, calcium salt 1:1 , dihydrate, brushite, aksepillen
PubChem CID 104805
ChEBI CHEBI:4496
IUPAC Name calcium;hydrogen phosphate;dihydrate
SMILES O.O.OP(=O)([O-])[O-].[Ca+2]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA40233A1
View Documents
Thermo Scientific Chemicals
040233A1
1 kg
Each for $164.70
Only null left
 
AA40233A7
View Documents
Thermo Scientific Chemicals
040233A7
5 kg
Each for $501.34
Only null left
 
Description

Description

Calcium hydrogen phosphate dihydrate is used in the manufacture of pharmaceutical tablets and as a source of calcium and phosphorus in nutritional supplements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7789-77-7
172.086
XAAHAAMILDNBPS-UHFFFAOYSA-L
104805
calcium;hydrogen phosphate;dihydrate
CaH5O6P
MFCD00149621
calcium hydrogenphosphate dihydrate, calcium hydrogen phosphate dihydrate, unii-o7tsz97gep, calcium monohydrogen phosphate dihydrate, o7tsz97gep, calcium phosphate dibasic dihydrate, dibasic calcium phosphate dihydrate, phosphoric acid, calcium salt 1:1 , dihydrate, brushite, aksepillen
CHEBI:4496
O.O.OP(=O)([O-])[O-].[Ca+2]

Specifications

109°C (in -H2O)
White
1 kg
CaHPO4·2H2O
14,1692
Slightly soluble in water; Soluble in dilute hydrochloric, nitric, and acetic acid; Insoluble in alcohol
O.O.OP(=O)([O-])[O-].[Ca+2]
172.086
CHEBI:4496
≤98%
2.306 g/mL
7789-77-7
Powder
CaH5O6P
MFCD00149621
calcium hydrogenphosphate dihydrate, calcium hydrogen phosphate dihydrate, unii-o7tsz97gep, calcium monohydrogen phosphate dihydrate, o7tsz97gep, calcium phosphate dibasic dihydrate, dibasic calcium phosphate dihydrate, phosphoric acid, calcium salt 1:1 , dihydrate, brushite, aksepillen
XAAHAAMILDNBPS-UHFFFAOYSA-L
calcium;hydrogen phosphate;dihydrate
104805
172.09 (136.06 Anhydrous)
Odorless
Calcium hydrogen phosphate dihydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 231-826-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only